VWR®, Sealing Film for Biological Cultures
Supplier: VWR Collection
VWRI731-0315EA 388 SGD
VWRI731-0315 VWRI731-0316 VWRI731-0313 VWRI731-0314
VWR®, Sealing Film for Biological Cultures
Seals Microplate Seals
Rayon film with medical grade adhesive. Hydrophobic, porous film in sheeted roll format for sealing tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth.
  • Recommended temperature range from −20 to +80 °C
  • Total thickness 139,7 μm
  • Pre-scored films separated by red colour bands for easy application
  • Pierceable and breathable
  • Pre-cut films on a roll
Delivery information: Each roll contains sufficient film to cover 50 plates.