VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWRI732-0548EA 73.5 SGD
VWRI732-0548
VWR® PCR Tubes, 0,2 ml
Tubes PCR Tubes
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.