VWR®, Sealing Mats for PCR Plates
Supplier: VWR Collection
VWRI732-0589EA 121 SGD
VWRI732-0589
VWR®, Sealing Mats for PCR Plates
Seals Microplate Seals
Transparent silicone mats fit most brands of 96-well PCR plates.
  • Sustainable design based on virgin resins
  • Can be cleaned and reused
  • Autoclavable at 121 °C
Designed to reduce evaporation when cycling 96-well plates.