VWR® Electroporation Cuvettes
Supplier: VWR Collection
VWRI732-1135EA 518 SGD
VWRI732-1135 VWRI732-1136 VWRI732-1137
VWR® Electroporation Cuvettes
Cuvettes Electroporation Cuvettes
These electroporation cuvettes are made from clear, medical grade polycarbonate and are compatible with all common commercially available electroporators.
  • Polished aluminium lowers arcing frequency
  • Colour-coded caps for easy identification of gap sizes
  • Round caps for easy, single-handed cap removal
Ordering information: Available in three gap sizes to meet common application requirements: 1 mm for highest field strength, suitable for bacteria; 2 mm gap for intermediate requirements; 4 mm gap for lowest field strength, suitable for mammalian cells andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable