VWR®, Filter Tips, Extra Long
Supplier: VWR Collection
VWRI732-1614EA 198 SGD
VWRI732-1614
VWR®, Filter Tips, Extra Long
Pipette Tips
Aerosol filter tips with extended length.
  • Sustainable design based on virgin resins
  • Supplied sterile, autoclavable at 122 °C
  • RNase-/DNase-, human DNA-, endotoxin-, protease-, ATP- and BPA-free
Delivery information: Supplied in hinged racks, racks are refillable.