VWR®, Cell Strainers
Supplier: VWR Collection
VWRI732-2757EA 117 SGD
VWRI732-2757 VWRI732-2758 VWRI732-2759
VWR®, Cell Strainers
Cell Strainers
Manufactured from a strong nylon mesh with evenly spaced mesh pores, these cell strainers are sterile, easy to use devices for quickly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Designed to protect valuable flow cytometry and cell sorting instrumentation by reliably removing clumps and debris from cell suspensions and clinical samples prior to analysis.
  • Available in three mesh sizes with different colours for easy identification
  • Extended lip on the strainer enables aseptic handling with forceps
  • Ready to
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M