VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWRI734-2328EA 347 SGD
VWRI734-2328 VWRI734-2323 VWRI734-2324 VWRI734-2325 VWRI734-2326 VWRI734-2327 VWRI732-2904 VWRI732-2906 VWRI732-2907 VWRI732-2905 VWRI734-2777 VWRI734-2778 VWRI734-2779 VWRI734-2780 VWRI734-2781 VWRI734-2782 VWRI734-2798 VWRI734-2799 VWRI734-2800 VWRI734-2801 VWRI734-2802 VWRI734-2803 VWRI734-2888
VWR® Multiwell Cell Culture Plates
Microplates Cell Culture Plates
VWR® multiwell cell culture plates are available with a choice of surface treatments.
  • Uniform well volume ensures an equal growth surface area
  • Well surface is smooth and free from striations to maximise usable growth area
  • Raised rims on wells with uniform rings on the lid to reduce evaporation
  • Single-position lid reduces the risk of cross-contamination and handling mistakes
  • Wells are labelled with alphanumeric code for easy identification
  • Suitable for use with all common instruments and automation
  • Sterilised by electron beam irradiation
  • Free from DNase and
More Product Information