PCR Workstation HEPA
Supplier: VWR Collection
VWRI732-3409EA 14870 SGD
VWRI732-3409 VWRI732-3410
PCR Workstation HEPA
Fume Hoods and Enclosures PCR Workstation Enclosures
The VWR® PCR Workstation HEPA is a PCR enclosure for PCR sample preparation with an HEPA Filter. It offers dual-decontamination action by UV inactivation of airborne and surface-bound contaminants, therefore, representing an ideal environment for PCR set up and other sensitive protocols. In addition, the HEPA filter with UV provides a barrier against dust, bacteria and mould.
  • Active decontamination of work surface during non working time by UV irradiation
  • Additional inactivation of aerosol-bound contaminants by shielded UV air recirculator during
More Product Information