VWR®, Universal Filter Tips
Supplier: VWR Collection
VWRI732-3633EA 250 SGD
VWRI732-3633 VWRI732-3630 VWRI732-3631 VWRI732-3632 VWRI732-3634 VWRI732-3636 VWRI732-3637 VWRI732-3638 VWRI732-3709
VWR®, Universal Filter Tips
Pipette Tips
VWR® Universal low retention filter tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with the widest range of pipette
  • Maximises sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Sterility Assurance Level (SAL) of 10-3
  • Autoclavable at 1 bar, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to 10 times moreMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing,Sustainable Packaging
Environmental certification FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic