VWR® Large Volume Cell Culture Plates
Supplier: VWR Collection
VWRI732-4428EA 0 SGD
VWRI732-4428
VWR® Large Volume Cell Culture Plates
Microplates Cell Culture Plates
6-well, round well plates for suspension cell culture.
  • Round well provides gentle mixing
  • Lower shearing force on cells
  • Compatible with laboratory automation
  • Virgin polystyrene construction
  • Height: 88 mm
  • Barcoded
The round well design is ideal for prokaryotic and eukaryotic cell culture. The round well provides an optimal swirling motion with low shear stress, preventing cell damage during incubation. Perfect for delicate cell lines such as CHO and hybridomas.
Packaging:
Plates are individually bagged with a lid.