VWR®, Drosophila Vials
Supplier: VWR Collection
VWRI734-1255EA 421 SGD
VWRI734-1255
VWR®, Drosophila Vials
Vials
These PS vials have a flat bottom straight wall design for convenient racking, stacking and filling.
The tray packed 10×10 row configuration is great for quick, effortless filling.
Bulk packed vials are layered with open-end orientation for convenient handling and smaller case size.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904