VWR®, Drosophila Stock Bottles
Supplier: VWR Collection
VWRI734-1249EA 884 SGD
VWRI734-1249 VWRI734-1250 VWRI734-1260 VWRI734-1261
VWR®, Drosophila Stock Bottles
Bottles Drosophila Bottles
PP, round or square bottom or PE, square bottom bottles.
  • Thin wall design for excellent visibility
  • Pliable top for easy removal of paper lid
  • Moulded-in food level graduation marks
Bottle 734-1259 features a wider edge for a more secure paper cap fit.
Bottle 734-1250 has a moulded indent near the bottom, which helps to secure media.
PP bottles are autoclavable, PE bottles are not.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable