VWR® Plugs for Drosophila Vials and Bottles
Supplier: VWR Collection
VWRI734-2672EA 409 SGD
VWRI734-2672 VWRI734-2673 VWRI734-2875
VWR® Plugs for Drosophila Vials and Bottles
Stoppers Cellulose Stoppers
These plugs are made of bonded dense weave cellulose acetate which provides an excellent barrier to mite infestation. They are precision manufactured for consistent fit and minimal waste.
  • Reduced evaporation, which keeps food fresh longer
  • Minimal shedding compared to cotton and rayon
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable