VWR®, Single-Use Tank Liners, Medical Grade
Supplier: VWR Collection
VWRI734-2943EA 2990 SGD
VWRI734-2943 VWRI734-2944 VWRI734-2945 VWRI734-2946 VWRI734-2947 VWRI734-2948 VWRI734-2949 VWRI734-2950 VWRI734-2951 VWRI734-2952 VWRI734-2953 VWRI734-2954
VWR®, Single-Use Tank Liners, Medical Grade
Bags Media Bags
These tank liners are single-use bags with an open top, designed to fit in cylindrical vessels.
  • No cleaning after media preparation
  • Medical grade, multi-layer film
  • For bioprocess applications
  • Delivered gamma irradiated and ready-to-use
These bags are used as a liner into a vessel to avoid cleaning of the vessel after media and buffer preparation. After installing the Tank Liner into a vessel and filling it with liquid, powder can be added to the vessel and the mixing process can start by using an overhead agitator. When mixing is finished, it’sMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable