VWR®, Single-Use Bioprocessing Bags
Supplier: VWR Collection
VWRI734-3304EA 3380 SGD
VWRI734-3304 VWRI734-3305 VWRI734-3306 VWRI734-3307 VWRI734-3308 VWRI734-3309 VWRI734-3310 VWRI734-3311 VWRI734-3312 VWRI734-3313 VWRI734-3314 VWRI734-3315 VWRI734-3316 VWRI734-3317 VWRI734-3318 VWRI734-3319 VWRI734-3320
VWR®, Single-Use Bioprocessing Bags
Bags Media Bags
Avantor single-use bioprocessing bags are designed to transfer and store your products throughout the drug manufacturing process. Made from proprietary FlexFilm™ material - a thick, co-extruded film featuring polyethylene inner and outer layers that provide extra resistance to a wide range of chemicals - Avantor single-use bags are supplied in a range of standard sizes to meet the needs of many bioprocessing applications.
  • Bag film made from proprietary ULDPE with high clarity and flexibility
  • Standard 2D bag assemblies provided at volumes ranging from 50 ml
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable