Avantor® Rigid Outer Support Containers
Supplier: VWR Collection
VWRI734-3421EA 5870 SGD
VWRI734-3421 VWRI734-3422 VWRI734-3423 VWRI734-3424 VWRI734-3425 VWRI734-3426 VWRI734-3427 VWRI734-3428 VWRI734-3429 VWRI734-3430 VWRI734-3431 VWRI734-3432
Avantor® Rigid Outer Support Containers
Bags Storage Bags
3D cubic bags can fit in existing support containers. These support containers are available in either High Density Polyethylene (HDPE) and Polypropelene (PP), or stainless steel material.
  • Standard 3D cubic bags are available in 100, 200, and 1000 L volumes with 3 top ports
  • Standard 3D cubic bags are available with or without a bottom port
  • Cleanroom and shipping variants are available in either material format
  • Plastic containers can collapse for easy storage
  • Stainless steel containers are ISTA certified for shipping
Avantor® 3D cubic bags are madeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable