Avantor® Magnetic Mixing System
Supplier: VWR Collection
VWRI734-3624EA 189670 SGD
VWRI734-3624 VWRI734-3629
Avantor® Magnetic Mixing System
Stirrers Overhead Stirrers
The Avantor Magnetic Mixing System is a robust, flexible, high-quality, expert designed solution for single use mixing needs.
  • Intuitive software: Controlled by a user-friendly software program that runs on a 7" colour touchscreen display. This software allows users to save mixing profiles and customise their mixing settings
  • Integrated loadcell: The system can have an integrated loadcell option that measures the weight of the vessel and contents, providing accurate feedback on the mixing process
  • Temperature controlled jacket: The system has a temperature
More Product Information