VWR®, Sterile Masks
Supplier: VWR Collection
VWRI76470-110EA 1030 SGD
VWRI76470-110
VWR®, Sterile Masks
Masks Hygiene Masks
VWR® made in USA sterile masks are an ideal option for single-use needs.
  • Individually bagged
  • Available with ear loop or tie configurations
  • 3-ply construction
  • Gamma irradiated to 10⁻⁶
  • Enclosed plastic encapsulated nose wire
A key part of the VWR® made in USA PPE line, these sterile cleanroom masks provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
Packaging: Each mask individually bagged sealed. 10 bags per poly pack. 20 packs per case. Case is poly lined.