VWR®, Sterile Alliance Sleeves
Supplier: VWR Collection
VWRI76491-706EA 2180 SGD
VWRI76491-706
VWR®, Sterile Alliance Sleeves
Disposable Garments
VWR® Made in USA sterile sleeves are an ideal option for single-use needs. Made with the specially formulated alliance breathable microporous fabric.
  • Individually bagged
  • Available with thumb loop configuration
  • ETO sterilised
  • Made with alliance breathable microporous fabric
  • Made in USA
A key part of the VWR® Made in USA PPE line, these sterile cleanroom sleeves provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
The Alliance fabric is designed to lower particulate levels by providingMore Product Information