VWR® Nano Spectrophotometer

Supplier: VWR Collection
VWRI76777-152EA 26320 SGD
VWRI76777-152
VWR® Nano Spectrophotometer
Spectrophotometers and Photometers Biophotometers
VWR® Nano Spectrophotometer is designed to simplify and accelerate your molecular biology research and routine applications.

  • Rapid calculation and display of 260/280 and 260/230 absorbance ratios
  • Extended-life xenon lamp and monochromator for full spectral scan
  • Auto adjusting pathlengths: 0,2 and 1,0 mm, ensuring accurate measurements
  • Detects up to 4500 ng/µl of undiluted dsDNA
  • Built-in printer and USB port
  • A cuvette port for optical density (OD 600) readings
  • Measurement and data processing time <5 sec
  • Pathlength control adjustment screw driven by
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Light source Pulsed xenon flash lamp
Detector 2048 Element linear silicone CCD array
Absorbance range Pedestal: 0,04 - 90 A, Cuvette: 0 - 4 A (10 mm equivalent)
Absorbance accuracy 1% (7,332 Abs at 260 nm)
Concentration Maximum: dsDNA: 4500 ng/µl, BSA: 135 mg/ml
Lower limit of detection: dsDNA: 2 ng/µl, BSA: 0,1 mg/ml
Output USB and printer
Sample size 0,5 - 2,0 µl
Wavelength accuracy ±1 nm
Electrical 240 V
Weight 2,8 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR