VWR®, Permanent Marker Pens
Supplier: VWR Collection
52877-810 52877-886
VWRI811-0055EA 78 SGD
VWRI811-0055 VWRI811-0056 VWRI811-0057 VWRI811-0058 VWRI811-0059 VWRU52877-810 VWRU52877-886
VWR®, Permanent Marker Pens
Pens Permanent Markers
Perfect for slides, sample bags and glassware.
  • Write on glass, plastic, porcelain, film, rubber, wood and paper
  • Permanent ink dries in seconds
Assorted colours: 6 black, 2 red, 2 blue, 2 green.