VWR®, Laboratory Markers
Supplier: VWR Collection
VWRI811-0068EA 107 SGD
VWRI811-0068 VWRI811-0066 VWRI811-0067
VWR®, Laboratory Markers
Pens Lab Markers
These versatile laboratory markers are designed to write on wet or dry surfaces, and are suitable for industrial, biomedical and educational laboratories.
  • Durable tip and generous ink supply ensure a long product life
  • Ink dries instantly, allowing immediate handling
  • Taper tip from 4mm to 1mm
  • Non sterile
  • Made without natural rubber latex
These markers provide permanent marking on plastic disposables. When used on glass, metal or porcelain surfaces, the ink will not smear when subjected to water, and can be washed off these surfaces using variousMore Product Information