VWR®, Labels, Cryo
Supplier: VWR Collection
VWRI817-5000EA 289 SGD
VWRI817-5000 VWRI817-5001 VWRI817-5003 VWRI817-5005 VWRI817-5006 VWRI817-5007 VWRI817-5008 VWRI817-5011 VWRI817-5013 VWRI817-5015
VWR®, Labels, Cryo
Labels
The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be used in most incubators.
  • No need to laminate
  • Use on tubes, racks and boxes