VWR® Duplicating Wire-O Laboratory Notebooks, Grid Format
Supplier: VWR Collection
VWRI818-0304EA 209 SGD
VWRI818-0304 VWRI818-0316 VWRI818-0317 VWRI818-0318 VWRI818-0319
VWR® Duplicating Wire-O Laboratory Notebooks, Grid Format
Notebooks
Wire-O laboratory notebooks feature a translux cover and grid format pages.
  • Translux cover
  • 215,9×279,4 mm pages
  • 50 duplicating pages
  • Grid format
Certifications: All books made in accordance with GLP, GMP, and GCP guidelines.