VWR® Wire-O Laboratory Notebooks, Grid Format
Supplier: VWR Collection
VWRI818-0303EA 82.2 SGD
VWRI818-0303 VWRI818-0312 VWRI818-0313 VWRI818-0314 VWRI818-0315
VWR® Wire-O Laboratory Notebooks, Grid Format
Notebooks
Wire-O laboratory notebooks feature a translux cover and grid format pages.
  • Translux cover
  • 215,9×279,4 mm pages
  • 50 pages
  • Grid format
Certifications: All books made in accordance with GLP, GMP, and GCP guidelines.