VWR® Duplicating Wire-O Laboratory Notebooks, Ruled Format
Supplier: VWR Collection
VWRI818-0305EA 200 SGD
VWRI818-0305 VWRI818-0320 VWRI818-0321 VWRI818-0322 VWRI818-0323
VWR® Duplicating Wire-O Laboratory Notebooks, Ruled Format
Notebooks
Wire-O laboratory notebooks feature a translux cover and ruled format pages.
  • Translux cover
  • 215,9×279,4 mm pages
  • 50 duplicating pages
  • Ruled format
Certifications: All books made in accordance with GLP, GMP, and GCP guidelines.