VWR® MX, Immersion Thermostat
Supplier: VWR Collection
VWRIMX-CV12EA 5420 SGD
VWRIMX-CV12
VWR® MX, Immersion Thermostat
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The MX immersion thermostat is user friendly with three control buttons and on-screen prompts. The convenient slide control enables the flow rate to be easily adjusted, and it can clamp securely to straight or curved tank walls.
  • Large EasyView™ LCD display with alarm or fault icons plus English prompts and menus
  • User-adjustable high and low temperature limits and low liquid level alarms
  • Single speed pressure pump
  • Integral pump and heater coil protection
  • Single point calibration
Maximum pump capacities:
11,9 l/min, 120 mbar.
Maximum pump capacities:
10More Product Information