VWR® Double Edge Blades, 10 Blade Dispenser
Supplier: VWR Products
AGBL-6007-0000
VWRMAGBL-6007-0000EA 7.3 SGD
VWRMAGBL-6007-0000
VWR® Double Edge Blades, 10 Blade Dispenser
Razors and Blades
Double edge blades feature a 3-Facet edge for exacting, clean cuts. Safety blade dispenser holds 10 blades.
  • Blade edge with MicroCoat technology enhances durability and sharpness
  • Facet edge for increased sharpness
The MicroCoat enrichment technology reduces cut force and adds a layer of protection for each blade helping the blade maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children's reach. Dispose of safely.
Order Now
SPECIFICATIONS
Environmentally Preferable