Unreinforced Silicone Tubing, Newage Industries
Supplier: VWR Products
APST-0250-0438
VWRMAPST-0250-0438EA 434 SGD
VWRMAPST-0250-0438
Unreinforced Silicone Tubing, Newage Industries
Tubing Transfer Tubing
Low-volatile grade, platinum-cured silicone tubing is clean-room produced for critical pharmaceutical, biomedical, cosmetic, and food applications. APST has undergone extensive physical, chemical, and biological testing.
  • Hardness value of 50 Shore A
  • Extremely flexible
  • Sterilisable by autoclave or gamma radiation
  • Imparts no taste or odor to critical streams
  • Manufactured and packaged in a Class 7 (Class 10000) ISO-certified clean room
  • Documented lot traceable with identification on bags
Resists temperature extremes, compression set, chemical attack,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable