VWR® Set of 5 Single Slits
Supplier: VWR Products
NP1000846
VWRMNP1000846EA 143 SGD
VWRMNP1000846
VWR® Set of 5 Single Slits
Educational Materials Physics Educational Materials Optics Learning Activities
Five single slits of different widths in a slide frame.
  • Slit widths: 0,1/ 0,2/ 0,4/ 0,8/ 1,6 mm
  • Dimensions: 50×50 mm²