VWR® Extractor™ Ethidium Bromide Waste Reduction System
Supplier: VWR Products
NP10448031
VWRMNP10448031EA 417 SGD
VWRMNP10448031
VWR® Extractor™ Ethidium Bromide Waste Reduction System
Extractors Liquid-Liquid Extractors
One-step filtration polypropylene funnel device for the rapid removal of ethidium bromide from gel-staining solutions.
It contains an activated carbon matrix, which removes >99% of ethidium bromide from electrophoretic buffer quickly and easily.
The extractor funnel device fits most standard laboratory flasks and bottles (neck size 33 to 45 mm), and the unit includes a cap for storage between uses. The polypropylene housing is chemically resistant to organics. Pack includes glass fiber prefilters, which remove gel pieces and other debris to avoid premature clogging of the carbonMore Product Information