VWR® Airstrip Rubber Mats, Black
Supplier: VWR Products
NP2423G
VWRMNP2423GEA 435 SGD
VWRMNP2423G
VWR® Airstrip Rubber Mats, Black
Floor Mats Traction and Anti-Fatigue Floor Mats
Made of 1,3 cm thick black rubber with air pockets underneath to give buoyancy.
Grease and chemical-resistant mats (NBR/SBR blend) also available.
Available as single sections (28×38") beveled on all sides to prevent tripping, or as interlocking sections (28×36") beveled on two sides. Two end sections interlock to make a 1,82 m length. Additional length is made up by center sections.
Order Now
SPECIFICATIONS
Environmentally Preferable