VWR® Cleanroom Hazmat Bags
Supplier: VWR Products
NP81167
VWRMNP81167EA 645 SGD
VWRMNP81167
VWR® Cleanroom Hazmat Bags
Bags Waste Disposal Bags
Durable LDPE bags are ideal for cleanroom use and disposing of hazardous materials.
  • Excellent clarity
  • Clear