Water, reagent

Supplier: VWR USA

Synonyms: Hydrogen oxide

NP9800-3
VWRMNP9800-3EA 162 SGD
VWRMNP9800-3
Water, reagent
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR