VWR® Personal Anti-Fatigue Mat™ ESD Insoles
Supplier: VWR Products
NPEPAMW89
VWRMNPEPAMW89EA 43.2 SGD
VWRMNPEPAMW89
VWR® Personal Anti-Fatigue Mat™ ESD Insoles
Shoe Insoles
MEGAComfort® Personal Anti-Fatigue Mat (PAM) ESD insoles are ergonomically designed by a doctor, using a patented combination of high-density, dual-layer 100% memory foam for anti-static footwear only. ESD threads provide tested electro-static dissipation. PAM ESD insoles do not make regular footwear ESD compliant.
  • ESD anti-static threads provide electro-static dissipation
  • Patented 100% dual-layer memory foam
  • Ergonomically designed by a doctor
  • Soft contoured arch support for balance, comfort, and cushioning
  • Silver ion anti-microbial technology in top cloth
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable