VWR® BioSorb® Biohazard Spill Control Powder
Supplier: VWR Products
NPINFBIO-20
VWRMNPINFBIO-20EA 2.9 SGD
VWRMNPINFBIO-20
VWR® BioSorb® Biohazard Spill Control Powder
Spill Control Materials
Super absorbent polymers solidify most liquids to gel form.
Ideal for use with any liquid specimen (infectious, diagnostic–Category B, or other). Packaged in a screw cap plastic bottle.
Order Now
SPECIFICATIONS
Environmentally Preferable