Glass jars for use in soil, sludge, and sediment sampling. Select sizes are for use in volatile organic analysis. See table for listings. I-CHEM Certified* 200 and 300 series bottles are processed to meet or exceed US EPA Specifications and Guidance 1 * 12 items
Supplier: VWR USA
Glass jars for use in soil, sludge, and sediment sampling. Select sizes are for use in volatile organic analysis. See table for listings. I-CHEM Certified* 200 and 300 series bottles are processed to meet or exceed US EPA Specifications and Guidance 1 * 12 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +65 6505 0760.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR at sales@sg.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.