VWR® Step-On Container
Supplier: VWR Products
NPRCP6146WHI
VWRMNPRCP6146WHIEA 407 SGD
VWRMNPRCP6146WHI
VWR® Step-On Container
Bins Waste Bins
Heavy-duty plastic waste container with overlapping lid raised by foot pedal. For hands-free sanitary waste management.
  • Easy-to-clean
  • Quiet operation
  • Resist chipping, rusting, and denting
  • Tight-fitting, overlapping lid contains odors
Quiet, heavy-duty pedal for hands-free sanitary use. Tough, puncture-resistant, all-plastic construction.
Container 56610-094 is supplied with a rigid, leakproof liner to contain liquids. A rigid liner for 68,1 L containers 66101-360 and -365 may be purchased separately. Mobile containers 19757-282 and 66101-361More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable