VWR® Calgonate® Emergency Hydrofluoric Acid Spill Kit
Supplier: VWR Products
NPTS044SPKIT01
VWRMNPTS044SPKIT01EA 871 SGD
VWRMNPTS044SPKIT01
VWR® Calgonate® Emergency Hydrofluoric Acid Spill Kit
Spill Control Materials
Calgonate® Hydrofluoric Acid bucket spill kits contain the personal protection equipment for users to safely neutralise and dispose of an hydrofluoric acid spill.
The 1-Bucket kits include a single 5-gallon bucket which contains personal protection equipment and supplies such as 1 tube of Calgonate® Calcium Gluconate gel (25 g), 1 bottle of Calgonate® Eyewash (4 oz.), and 4 lbs. of neutraliser. The 2-Bucket kit consists of 2 bright 5-gallon buckets, containing personal protection equipment for 2 people and the products to neutralise and clean up hydrofluoric acid spills.