VWR® Ultra high vacuum aluminium foil
Supplier: VWR Products
NPX11220118
VWRMNPX11220118EA 718 SGD
VWRMNPX11220118
VWR® Ultra high vacuum aluminium foil
Aluminium Foils
Designed for use in controlled environments, ultra high vacuum aluminium foil is virtually free from contamination and residual rolling oils. Foil is available in several sizes and supplied on plastic cores. For extra protection against contamination, it is packaged in a plastic dispenser box and wrapped in film.