VWR® Supelco® SpillSolv® Laboratory Spill Kit
Supplier: VWR Products
OTEM-SX1300-1
VWRMOTEM-SX1300-1EA 1780 SGD
VWRMOTEM-SX1300-1
VWR® Supelco® SpillSolv® Laboratory Spill Kit
Spill Control Materials
For safe, easy, effective spill treatment in the laboratory.
  • Agents are formulated to rapidly clean up chemical spills and vapors
  • Each bottle has a convenient dispensing cap for shaking or pouring
Caustics and acids are converted into nonhazardous compounds that meet EPA specifications for nonhazardous waste, simplifying cleanup and disposal. Solvent spills can be easily treated and transported with the absorbent in the solvent spill kit. This absorbent will also act as a vapor suppressant. The formaldehyde spill kit contains a two-part neutralisationMore Product Information