VWR® Q Path®, Slow Acting Decalcifiers
Supplier: VWR Q-Path Chemicals
VWRQ11028305EA 30.8 SGD
VWRQ11028305
VWR® Q Path®, Slow Acting Decalcifiers
Stains Microscopy and staining reagents
DC1 slow acting decalcifiers contain a mixture of formic acid and formalin.
  • Decalcify and fix small fragments (5 mm) in two to four hours
  • Recommended volume is 20× the fragment volume
Ready to use for applications in histology and pathology laboratories.