VWR®, Freestanding and Mobile Glassware Washer
Supplier: VWR Collection
10002-852
VWRU10002-852EA 19540 SGD
VWRU10002-852
VWR®, Freestanding and Mobile Glassware Washer
Glassware Washing Machines
Engineered for durable, reliable and energy efficient operation, these Glassware Washers can be either Freestanding or Mobile and are supplied with mounting feet and wheels.
  • 304 stainless steel washing chamber and full exterior casing ensures long life and corrosion protection plus maintains superior cosmetic appearance
  • Three separate filters protect pumps from debris and are conveniently located for easy access
  • Multiple cycle selections and microprocessor control systems ensure that each load is processed efficiently with minimum energy consumption
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Electrical 115 V, 60 Hz, 15 A
W×D×H 596×550×978 mm