VWR®, Microcentrifuge tubes, with screw cap, standard line
Supplier: VWR Collection
10025-756
VWRU10025-756EA 92.3 SGD
VWRU10025-756
VWR®, Microcentrifuge tubes, with screw cap, standard line
Tubes Microcentrifuge Tubes
These are crystal-clear PP tubes that are available in conical bottom and self-standing bottom formats and ideal for sample processing applications.
  • Tubes can be spun up to 20000 ×g
  • Autoclavable and freezable
  • DNase-free, RNase-free and non-pyrogenic
Certifications: TSE/BSE-free certification.
Ordering information: Caps and color inserts are has to be ordered seperately.
Packaging: Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Sterile tubes are sold with attached cap and non sterile tubes are sold without cap.
Order Now
SPECIFICATIONS
Environmentally Preferable