VWR® FILTRA-BAG®, Blender bags
Supplier: VWR Collection
10048-882
VWRU10048-882EA 234 SGD
VWRU10048-882
VWR® FILTRA-BAG®, Blender bags
Bags Blender Bags
These FILTRA-BAG® blender bags are designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances.
  • Made with a PE/PET outer shell which practically eliminates the possibility of piercing the bag wall during blending
  • Prevents pipette blockage as well as uneven plate distribution
  • Dividing filter membrane has 1840 holes per square inch with a pore size of approximately 330 microns