VWR®, Mini Incubator, Digital
Supplier: VWR Collection
10054-476
VWRU10054-476EA 1830 SGD
VWRU10054-476
VWR®, Mini Incubator, Digital
Incubators Compact Benchtop Incubators
These digital incubator provides an economical and space saving solution for microbiology or haematology applications.
  • Digital controls
  • Personal sized
The temperature can be freely adjusted in increments of 0,1 °C up to a maximum of 70 °C. Heating elements are incorporated into the housing base and walls to ensure temperature stability and uniformity. The door of the incubator has a transparent plexiglas® window to provide full visibility to samples. The housing and door frame are constructed entirely of epoxy coated mild steel for strength and durability.