VWR®, Cell Scrapers
Supplier: VWR Collection
10062-904
VWRU10062-904EA 176 SGD
VWRU10062-904
VWR®, Cell Scrapers
Cell Scrapers
Specifically designed to make collecting of cells from flasks easier and more effective.
  • Cross-ribbed handle in ABS provides a greater rigidity and ensures a better control while scraping cells
  • Thin, flexible TPE blade prevents damage to cells
  • Individually wrapped
  • Sterilised by gamma irradiation
  • Non pyrogenic