VWR®, Erlenmeyer Flasks, Wide Neck, with Screw Cap
Supplier: VWR Collection
89095-274 89095-266 89095-282 89095-270 10126-412
VWRU89095-274EA 780 SGD
VWRU89095-274 VWRU89095-266 VWRU89095-282 VWRU89095-270 VWRU10126-412
VWR®, Erlenmeyer Flasks, Wide Neck, with Screw Cap
Flasks Erlenmeyer Flasks
PC, clear, graduated.
  • Molded-in-graduations for accuracy
  • RNase- and DNase-free
  • Sterilised by gamma radiation
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
These Erlenmeyer culture flasks are disposable and ideal for all shaker culture applications as well as liquid handling and storage.
Sterility Assurance Level (SAL) of 10⁻⁶ and nonpyrogenic certification.
Order Now
SPECIFICATIONS
Environmentally Preferable