VWR®, Tube Rotator
Supplier: VWR Collection
10136-084
VWRI444-0500EA 669 SGD
VWRI444-0500 VWRI444-0501 VWRI444-0502 VWRU10136-084
VWR®, Tube Rotator
Shakers Rotating/Rocking Shakers
A unique design of rotator makes this unit suitable for a wide range of mixing applications in biochemistry, molecular biology and histochemistry. The rotisserie assemblies allow for mixing tubes vertically, horizontally, or any position in between, therefore, gentle mixing, using a horizontal position is easily accommodated for applications such as hybridisation. Alternatively rotation with vertically positioned tubes produces a tumbling motion suitable for mixing samples in blood tubes. The two halves of the rotisserie can be adjusted independently of each other to increase versatility andMore Product Information