VWR®, Plate Centrifuge
Supplier: VWR Collection
10144-214
VWRU10144-214EA 2820 SGD
VWRU10144-214
VWR®, Plate Centrifuge
Centrifuges Microcentrifuges
Spins at 2500 rpm/ 850 ×g, to quickly spin down droplets and condensation in PCR plates.
  • Easy access rotor includes two plate carriers
  • Unique swing-out rotor design prevents sample spillage
  • Less than half the size of traditional centrifuges
Accepts all popular PCR plates: skirted, half-skirted and no-skirted. Grey colour.
Ordering information: Two pairs of adapter plates included. Two-year parts and labour warranty.